Sodium carbonate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Calcined soda, Carbonic acid disodium salt
436800050 436800250
ACRO436800050EA 46.1 EUR
ACRO436800050 ACRO436800250
Sodium carbonate 99.999% (trace metals basis)
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (760 mmHg)
Melting Pt: 851 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder
Total trace metal impurities ≤10 ppm