Tetraammineplatinum(II) chloride hydrate 99.995% (trace metals basis)
Supplier: Thermo Scientific
Warning
Synonyms: AKOS015924294, AC-24209
437000050 437005000
ACRO437000050EA 829 EUR
ACRO437000050 ACRO437005000
Tetraammineplatinum(II) chloride hydrate 99.995% (trace metals basis)
Tetraammineplatinum(II) chloride hydrate
Formula: Pt(NH₃)₄Cl₂·xH₂O
MW: 334.11 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149947
CAS Number: 108374-32-9
Order Now
Specification Test Results
Appearance Off-white to light yellow Crystalline powder
Total trace metal impurities ≤50 ppm
Platinum (Pt) 51 to 59 % (typical)