3-Bromo-5-chloropyridine 97%
Supplier: Thermo Scientific
Danger
437400050 437400250
ACRO437400050EA 140 EUR
ACRO437400050 ACRO437400250
3-Bromo-5-chloropyridine 97%
3-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Melting Pt: 78.7…80.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 77.0 to 81.0 °C
Water ≤3.00 %
GC ≥96.0 %