3-Bromo-5-fluoropyridine 97%
Supplier: Thermo Fisher Scientific
Danger
437420010 437420050
ACRO437420010EA 39.4 EUR
ACRO437420010 ACRO437420050
3-Bromo-5-fluoropyridine 97%
3-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 78 °C (11Torr)
Melting Pt: 24…28 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD04112555
CAS Number: 407-20-5
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Colorless to white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %