6-Chloro-7-deazapurine 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine
437440010 437440050
ACRO437440010EA 93.2 EUR
ACRO437440010 ACRO437440050
6-Chloro-7-deazapurine 97%
6-Chloro-7-deazapurine
Formula: C₆H₄ClN₃
MW: 153.57 g/mol
Melting Pt: 180.9…182.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01686865
CAS Number: 3680-69-1
UN: 2811
Order Now
Specification Test Results
Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %