4-Cyanophenylacetic acid 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-(4-Cyanophenyl)acetic acid
437730010 437730050
ACRO437730010EA 120 EUR
ACRO437730010 ACRO437730050
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid
Formula: C₉H₇NO₂
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06798066
CAS Number: 5462-71-5
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150.0 to 155.0 °C
HPLC ≥96.0 %