Ethynylmagnesium bromide 0.5 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Magnesium, bromoethynyl-, Bromo(ethynyl)magnesium
439121000 439128000
439121000.EA 52.1 EUR
439121000. 439128000.
Ethynylmagnesium bromide 0.5 M in THF, AcroSeal®
Ethynylmagnesium bromide
Formula: C₂HBrMg
Density: 0.94 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00075342
CAS Number: 4301-14-8
UN: 3399
ADR: 4.3,II
Order Now
Specification Test Results
Appearance Colorless to yellow to grey to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M