1,3-Benzothiazole-6-carboxylic acid 96%
Supplier: Thermo Scientific
Warning
Synonyms: Benzo[d]thiazole-6-carboxylic acid, Benzothiazole-6-carboxylic acid
439700050 439700250
ACRO439700050EA 79.5 EUR
ACRO439700050 ACRO439700250
1,3-Benzothiazole-6-carboxylic acid 96%
1,3-Benzothiazole-6-carboxylic acid
Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Melting Pt: 241…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00111651
CAS Number: 3622-35-3
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point ≥240.0 °C (Dec)
HPLC ≥95.0 %