2-Chlorophenethyl bromide 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
440140010
440140050
ACRO440140010EA
34.5
EUR
ACRO440140010
ACRO440140050
2-Chlorophenethyl bromide 95%
2-Chlorophenethyl bromide
Formula:
C₈H₈BrCl Boiling Pt: 110…112 °C (10Torr) Density: 1.4569 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04117464 CAS Number: 16793-91-2 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |