3-Fluoro-4-hydroxybenzoic acid 95%
Supplier: Thermo Fisher Scientific
Warning
441890010 441890050
ACRO441890010EA 56.9 EUR
ACRO441890010 ACRO441890050
3-Fluoro-4-hydroxybenzoic acid 95%
3-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00143161
CAS Number: 350-29-8
Order Now
Specification Test Results
Appearance Off-white to brown Powder
Infrared spectrum Conforms
HPLC ≥94.0 %