D-α-Aminoadipic acid 98%
Supplier: Thermo Scientific
ACRO441960050EA 905 EUR
ACRO441960050
D-α-Aminoadipic acid 98%
D-α-Aminoadipic acid
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 43.60 to 45.84 % (C)
Elemental Analysis 8.47 to 8.91 % (N)
Specific optical rotation -17° to -23° (20°C, 589 nm) (c=2 in 6N HCl)