L-Alanine-tert-butyl ester hydrochloride 99%
Supplier: Thermo Fisher Scientific
Synonyms: L-Alanine tert-Butyl Ester Hydrochloride, tert-Butyl (S)-2-Aminopropionate Hydrochloride, H-Ala-OtBu·HCl, (S)-2-Aminopropionic acid tert-Butyl Ester Hydrochloride, H-Ala-OtBu.HCl, (S)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride
442920010 442920050
ACRO442920010EA 30.6 EUR
ACRO442920010 ACRO442920050
L-Alanine-tert-butyl ester hydrochloride 99%
L-Alanine-tert-butyl ester hydrochloride
Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 206…216 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035524
CAS Number: 13404-22-3
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥98.5 to ≤101.5 %
Specific optical rotation +1.2° to +1.6° (20°C, 589 nm) (c=2 in EtOH)