4-Chloro-3-fluoropyridine 95%
Supplier: Thermo Fisher Scientific
Warning
443160050 443160250
ACRO443160050EA 89.6 EUR
ACRO443160050 ACRO443160250
4-Chloro-3-fluoropyridine 95%
4-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 142…143 °C (760 mmHg)
Melting Pt: 27…24 °C
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %