3-Iodo-1H-pyrazole 97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Iodopyrazole
444810010 444810050
ACRO444810010EA 119 EUR
ACRO444810010 ACRO444810050
3-Iodo-1H-pyrazole 97%
3-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.98 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4
Order Now
Specification Test Results
Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 67 to 74 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure