2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Warning
444920010 444920050
ACRO444920010EA 168 EUR
ACRO444920010 ACRO444920050
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 97%
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄
MW: 265.12 g/mol
Melting Pt: 39…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788240
CAS Number: 214360-59-5
Order Now
Specification Test Results
Appearance Colorless to off-white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure