tert-Butyldimethylsilane 95%
Supplier: Thermo Fisher Scientific
Danger
445710050 445710250
ACRO445710050EA 54.1 EUR
ACRO445710050 ACRO445710250
tert-Butyldimethylsilane 95%
tert-Butyldimethylsilane
Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 81…83 °C
Density: 0.701 g/cm³
Flash Pt: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010754
CAS Number: 29681-57-0
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94.0 %