Cyclopropylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
445951000 445958000
445951000.EA 235 EUR
445951000. 445958000.
Cyclopropylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Cyclopropylmagnesium bromide
Formula: C₃H₅BrMg
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00672094
CAS Number: 23719-80-4
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Colorless to yellow to orange Solution
Titration Grignard 0.40 to 0.60 M