Methylmagnesium bromide 1 M in CPME (cyclopentyl methyl ether), AcroSeal®
Supplier: Thermo Scientific
Danger
446001000 446008000
446001000.EA 70.1 EUR
446001000. 446008000.
Methylmagnesium bromide 1 M in CPME (cyclopentyl methyl ether), AcroSeal®
Methylmagnesium bromide
Formula: CH₃BrMg
MW: 119.24 g/mol
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000041
CAS Number: 75-16-1
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Dark grey Solution (May develop some turbidity or precipitate)
Titration Grignard 0.95 to 1.10 M