Triisopropylsilane 98%, AcroSeal®

Supplier: Thermo Scientific
Warning

446171000.EA 123 EUR
446171000.
Triisopropylsilane 98%, AcroSeal®
Triisopropylsilane
Formula: C₉H₂₂Si
MW: 158,36 g/mol
Boiling Pt: 84…86 °C (35 mmHg)
Density: 0,773 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009657
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR