Potassium permanganate ACS Hg_low
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Permanganic acid potassium salt
447211000 447215000
ACRO447211000EA 27.9 EUR
ACRO447211000 ACRO447215000
Potassium permanganate ACS Hg_low
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824
Order Now
Specification Test Results
Appearance Dark green to brown-black Crystalline powder
Titration with KMnO4 ≥99.0 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO4) ≤0.02 %
Mercury (Hg) ≤0.05 ppm
Insoluble matter ≤0.2 %