Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (A-taPhos)2PdCl2, PdCl2(Amphos)2, Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II), Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
447890010 447890050
447890010.EA 155 EUR
447890010. 447890050.
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 95%
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
Formula: C₃₂H₅₆Cl₂N₂P₂Pd
MW: 708.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265123
CAS Number: 887919-35-9
Order Now
Specification Test Results
Appearance Yellow to dark yellow to dark orange Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥94 %