Ammonium molybdate(VI) 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate
448540050 448540250 448541000
ACRO448540050EA 123 EUR
ACRO448540050 ACRO448540250 ACRO448541000
Ammonium molybdate(VI) 99.99% (trace metals basis)
Ammonium molybdate(VI)
Formula: H₈MoN₂O₄
MW: 196.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011596
CAS Number: 13106-76-8
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals and/or chunks
Total trace metal impurities ≤100 ppm
Molybdenum (Mo) 47.0 to 50.7 %