2-Fluoro-4-pyridineboronic acid 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid
448730010 448730050
ACRO448730010EA 136 EUR
ACRO448730010 ACRO448730050
2-Fluoro-4-pyridineboronic acid 98%
2-Fluoro-4-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD04112534
CAS Number: 401815-98-3
Order Now
Specification Test Results
Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %