Tris(2-cyanoethyl)phosphine 95%
Supplier: Thermo Scientific
Danger
Synonyms: Tris (2-cyanoethyl)phosphine, 3,3',3''-Phosphinetriyltris(propiononitrile), 3,3',3''-Phosphinidynetris(propiononitrile)
449710010 449710100 449710500
449710010.EA 25.1 EUR
449710010. 449710100. 449710500.
Tris(2-cyanoethyl)phosphine 95%
Tris(2-cyanoethyl)phosphine
Formula: C₉H₁₂N₃P
MW: 193.19 g/mol
Boiling Pt: 235 °C (.9 mmHg)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00013828
CAS Number: 4023-53-4
UN: 3464
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 99 °C
Titration Iodimetric ≥94 %