Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific
Danger
450580025 450580100 450580250
ACRO450580025EA 53.4 EUR
ACRO450580025 ACRO450580100 ACRO450580250
Sodium hydroxide 98%, flakes, pure
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Appearance White Flakes
Titration with HCl ≥97.5 %