2-Fluoro-5-formylpyridine 95%
Supplier: Thermo Scientific
Warning
Synonyms: 6-Fluoropyridine-3-carboxaldehyde, 6-Fluoronicotinaldehyde
449850010 449850050 449850250
ACRO449850010EA 52.7 EUR
ACRO449850010 ACRO449850050 ACRO449850250
2-Fluoro-5-formylpyridine 95%
2-Fluoro-5-formylpyridine
Formula: C₆H₄FNO
MW: 125.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064049
CAS Number: 677728-92-6
Order Now
Specification Test Results
Appearance Colorless to white to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94 %