tert-Butyl-3-aminoazetidine-1-carboxylate 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-Amino-1-Boc-azetidine, tert-Butyl 3-aminoazetidine-1-carboxylate, 1-BOC-3-aminoazetidine
451180010 451180050
ACRO451180010EA 106 EUR
ACRO451180010 ACRO451180050
tert-Butyl-3-aminoazetidine-1-carboxylate 95%
tert-Butyl-3-aminoazetidine-1-carboxylate
Formula: C₈H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01861753
CAS Number: 193269-78-2
UN: 2735
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %