1,3-benzothiazol-5-amine 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole
451540050 451540250
ACRO451540050EA 465 EUR
ACRO451540050 ACRO451540250
1,3-benzothiazol-5-amine 95%
1,3-benzothiazol-5-amine
Formula: C₇H₆N₂S
MW: 150.199 g/mol
Melting Pt: 73…77.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115282
CAS Number: 1123-93-9
Order Now
Specification Test Results
Appearance White to yellow Powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %