Imidazo[1,2-a]pyridine-6-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Warning
451740010 451740100
ACRO451740010EA 56.5 EUR
ACRO451740010 ACRO451740100
Imidazo[1,2-a]pyridine-6-carboxylic acid 97%
Imidazo[1,2-a]pyridine-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07021498
CAS Number: 139022-25-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %