3,5-Difluoro-4-pyridinamine 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Amino-3,5-difluoropyridine, 3,5-Difluoropyridin-4-amine
452530010 452530050
ACRO452530010EA 163 EUR
ACRO452530010 ACRO452530050
3,5-Difluoro-4-pyridinamine 97%
3,5-Difluoro-4-pyridinamine
Formula: C₅H₄F₂N₂
MW: 130.1 g/mol
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD01862101
CAS Number: 159783-22-9
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 100 to 105 °C
GC ≥96.0 %