1,3,5-Trifluoro-2,4,6-triiodobenzene 97%
Supplier: Thermo Scientific
Warning
452690010 452690050
ACRO452690010EA 99.8 EUR
ACRO452690010 ACRO452690050
1,3,5-Trifluoro-2,4,6-triiodobenzene 97%
1,3,5-Trifluoro-2,4,6-triiodobenzene
Formula: C₆F₃I₃
MW: 509.77 g/mol
Boiling Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD06248899
CAS Number: 84322-56-5
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 159 °C
GC ≥96.0 %