Cobalt 99+%, powder, pure
Supplier: Thermo Fisher Scientific
Danger
453910010 453912500
ACRO453910010EA 516 EUR
ACRO453910010 ACRO453912500
Cobalt 99+%, powder, pure
Cobalt
Formula: Co
Boiling Pt: 2870 °C (760 mmHg)
Melting Pt: 1495 °C
Density: 8,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Grey Powder
Titration Complexometric ≥99.0 %