Bismuth(III) nitrate pentahydrate 98%, pure
Supplier: Thermo Scientific
Danger
Synonyms: Bismuth trinitrate pentahydrate
454110010 454110050
ACRO454110010EA 136 EUR
ACRO454110010 ACRO454110050
Bismuth(III) nitrate pentahydrate 98%, pure
Bismuth (III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Density: 2,736 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
UN: 1477
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystals
Titration Complexometric ≥97.5 %