2-Fluoro-α-methyl-4-biphenylacetic acid 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
454660250 454660010 454660050
ACRO454660250EA 269 EUR
ACRO454660250 ACRO454660010 ACRO454660050
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid
Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 110 to 116 °C
HPLC ≥97.5 %