2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%
Supplier: Thermo Scientific
Danger
Synonyms: 2,2,2-Trifluoroethyltrifluoromethanesulphonate
454970010 454970050 454970250
ACRO454970010EA 44.9 EUR
ACRO454970010 ACRO454970050 ACRO454970250
2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Formula: C₃H₂F₆O₃S
MW: 232.10 g/mol
Boiling Pt: 89…91 °C (740 mmHg)
Density: 1.61 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00671579
CAS Number: 6226-25-1
UN: 2927
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %