Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
93-260/9.5-10 93-260/6.5-7 93-260/7.5-8 93-260/8.5-9 93-260/10.5-11 93-260/5.5-6
112-4642EA 41.1 EUR
112-4642 112-4639 112-4640 112-4641 112-4637 112-4638
Gloves, nitrile/neoprene, Microflex® 93-260
Gloves
These disposable heavyweight gloves feature a three layer design for superior protection against harsh chemicals including acids, solvents and bases.
  • Powder-free, textured fingertips
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
  • Lower acceptable pinhole rate (0,65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone-free formulation and processing ensure better product protection
Ideal for blendingMore Product Information
Order Now
SPECIFICATIONS
Model 93-260
Ambidextrous / hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex / latex-free Latex-free
Powdered / powder-free Powder-free
Finish Textured fingertips
Cuff style Beaded
Finger thickness 0,20 mm
Palm thickness 0,19 mm
EN norm EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS, EN 421, EN 388:2016 (2000X)
Anti-static +