Cut resistant gloves, Check & Go Amber PU 3
Supplier: HONEYWELL SAFETY
2332242/10 2332242/11 2332242/7 2332242/8 2332242/9
112-4643EA 0 EUR
112-4643 112-4644 112-4645 112-4646 112-4647
Cut resistant gloves, Check & Go Amber PU 3
Gloves
These polyamide/HPPE fibres/elastane knitted gloves with PU coating on palm and fingertips provide cut and puncture resistance. Ideal for use when handling sharp parts in a dry or greasy environment.
  • Knitting technique helps produce a glove that fits the hands snugly, with good dexterity
  • Tight knit (gauge 13) gives the glove perfect support, limits penetration of dirt and offers good sense of touch
  • Ventilated back optimises breathability
  • PU coating of the palm and fingertips provides outstanding abrasion resistance
  • Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise the level of cut protection
Typical application areas include: Automotive and part manufacturing, aviation building and construction, catering, energy or electricity, glass industries, metal and paper industries, logistics and maintenance.
Caution: Do not expose this glove to the heat.
Certifikace: EN 388 (4342); Cat. II
Upozornení: Please note: Do not expose this glove to heat.