ESD safety boots, lace-up, Safety-Grip, Renzo S High
Supplier: ELTEN
764751/45 764751/46 764751/47 764751/48 764751/49 764751/50 764751/36 764751/37 764751/38 764751/39 764751/40 764751/41 764751/42 764751/43 764751/44
114-7235EA 93.9 EUR
114-7235 114-7236 114-7237 114-7238 114-7239 114-7240 114-7226 114-7227 114-7228 114-7229 114-7230 114-7231 114-7232 114-7233 114-7234
ESD safety boots, lace-up, Safety-Grip, Renzo S High
Shoes and Boots
Durable ESD safety boots made from robust, breathable cowhide leather, a full padded bellows tongue and a dual density rubber/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip plus good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Sole matches the movement of the foot, for more stability and less fatigue
  • Heat resistant seams and laces
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, HI, HRO.