Polyvinyl alcohol, M.W. 500 - 5,000 85-89 %, high purity, hydrolysed
Supplier: VWR Chemicals
Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®
9664-100GEA 102 EUR
9664-100G
Polyvinyl alcohol, M.W. 500 - 5,000 85-89 %, high purity, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5
Order Now
Specification Test Results
Degree of Hydrolysis 85 – 89%
Loss on Drying ≤5.0%
pH (4.0%, Water) @25 °C 4.5 – 7.0
Residue after Ignition ≤2.0%
Viscosity Passes Test