XTT sodium salt, high purity

Supplier: VWR Chemicals

N812-100MGEA 0 EUR
N812-100MG N812-5G
XTT sodium salt, high purity
XTT sodium salt
Formula: C₂₂H₁₇N₇O₁₃S₂Na
MW: 673,52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083517
CAS Number: 111072-31-2

Order Now

Specification Test Results

Emax (285 nm) 14800 – 16800
Appearance Yellow powder
Water (KF) ≤15%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR