Baumé hydrometers with thermometer
Supplier: Amarell
AMARH843210-FLEA 71.45 EUR
AMARH843210-FL AMARH843216-FL AMARH843290-FL AMARH843288-FL
Baumé hydrometers with thermometer
Hydrometers
Baumé hydrometers with thermometer calibrated at 15 °C.
  • Red or blue filing liquid
  • Round capillary
  • Accuracy: ±1 °C
These paper scale thermometers are designed with well visible print, shot weighted body, at the bottom pointed end. Thermometer is presented inside the hydrometer body.