VWR®, Safety Caps
Supplier: VWR
VWR®
590-0038EA 107 EUR
590-0038 590-0036 590-0037
VWR®, Safety Caps
Closures Safety Caps and Safety Waste Caps
Extract solvents or mobile phases safely from reservoirs and bottles, without being exposed to hazardous gases and solvent vapours. High quality materials (PTFE and PP) ensure maximum safety for your laboratory staff and environment.
  • Recommended for HPLC systems
  • Chemically resistant against aggressive organic solvents
  • Air valve should be changed every 6 months for maximum safety
Delivery information: Supplied with an air valve/filter and corresponding fitting.