Sodium dodecylbenzenesulphonate 80, flakes_off-white with orangish hue
Supplier: MP Biomedicals
Danger
Synonyms: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate
0215788983 0215788991
ICNA0215788983EA 36 EUR
ICNA0215788983 ICNA0215788991
Sodium dodecylbenzenesulphonate 80, flakes_off-white with orangish hue
Sodium dodecylbenzenesulphonate
Dodecylbenzene Sulfonic Acid is a member of the linear alkylbenzenesulfonates.
  • Off-white fakes with orangish hue
Sodium dodecylbenzenesulfonate was used to fabricate amperometic biosensors for glucose.
Sodium dodecylbenzenesulfonate (SDBS) acts as a surfactant in the ionic self-assembly reaction of cyclophane BIMCP-1.
Soluble in 10% aqueous clear, slightly yellow solution.
Formula: C₁₈H₂₉NaO₃S
MW: 348.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011508
CAS Number: 25155-30-0
UN: 3077
ADR: 9 III
Merck Index: 13,08686
Order Now
Specification Test Results
Identity Test Passes
Purity ≥80%
FTIR Conforms to Reference