DL-α-Tocopherol acetate, powder
Supplier: MP Biomedicals
Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
0210055525 0210055590
ICNA0210055525EA 54.9 EUR
ICNA0210055525 ICNA0210055590
DL-α-Tocopherol acetate, powder
DL-α-Tocopherol acetate
D-α-tocopherol is the predominant form of vitamin E in plasma and tissues. This product is a semi-synthetic acetate ester of naturally occuring α-tocopherol.
  • Presentation: Powder
  • Biochemical Category: Lipids and Vitamins
α-Tocopherol is a powerful inhibitor of the proliferation of estrogen receptor positive and estrogen receptor negative human breast cancer cell lines in a dose dependent manner in vitro.
Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Tocopherol acetate has properties similar but not identical to α-tocopherol.
Cut with corn starch and hydrolyzed protein.
Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
MDL Number: MFCD00072042
CAS Number: 7695-91-2
Order Now
Specification Test Results
Identity Test Passes
Activity 250 u/g