Periodic acid ≥99%, pale yellow crystalline powder

Supplier: MP Biomedicals
Danger

ICNA0210259580EA 80.6 EUR
ICNA0210259580
Periodic acid ≥99%, pale yellow crystalline powder
Periodic acid

Periodic acid is a compound used to stain the cytoplasm of cells found in the tubular pattern, mucus, and apical surfaces of cells of various lumen and glycoproteins on SDS-PAGE gels.


Periodic acid is used for staining of cell cytoplasm . Also periodic acid can be used in oxidation reactions converting sulphides into sulphones, catalysed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones. Periodic acid can also react with the 1,2 glycol linkage of carbohydrates forming aldehydes in tissue sections which can be colored with Schiff bases, thus allowing for the staining of these tissues. Furthermore, studies show that periodic acid is not effective in staining cancer cells in solid nests, which are better stained using Grimelius .


Freely soluble in water (100 mg/ml), alcohol, nitric acid; slightly soluble in ether.

Formula: H₅IO₆
MW: 227,94 g/mol
MDL Number: MFCD00011344
CAS Number: 10450-60-9
UN: 3085
ADR: 5.1,II
https://si.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Specification Test Results

Form Yellowish Crystalline Powder
Storage Store at Room Temperature (15 - 30 °C). Store Desiccated.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR