Reactive red 4, powder C.I. 18105
Supplier: MP Biomedicals
Synonyms: tetrapotassium 5-benzamido-3-{2-[5-({4-chloro-6-[(4-sulphonatophenyl)amino]-1,3,5-triazin-2-yl}amino)-2-sulphonatophenyl]diazen-1-yl}-4-hydroxynaphthalene-2,7-disulphonate, Procion Reactive Red 4, Cibacron Brilliant Red 3B-A
0215201705 0215201725
ICNA0215201705EA 73 EUR
ICNA0215201705 ICNA0215201725
Reactive red 4, powder C.I. 18105
Reactive red 4
A monochlorotriazine dye that inhibits phosphotransferase acitvity
Soluble in water. Very slightly soluble in ethanol and methanol.
Formula: C₃₂H₁₉ClN₈O₁₄S₄Na₄
MW: 995.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009690
CAS Number: 17681-50-4
Order Now
Specification Test Results
Identity Test Passes
λmax (ethanol) 282 ±2 nm