Copper(II) nitrate trihydrate, powder
Supplier: MP Biomedicals
Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate
0215350180 0215350190
ICNA0215350180EA 34.5 EUR
ICNA0215350180 ICNA0215350190
Copper(II) nitrate trihydrate, powder
Copper (II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114
MDL Number: MFCD00149671
CAS Number: 10031-43-3
UN: 1477
ADR: 5.1 II
Merck Index: 13,02671