Longifolene, clear, colourless liquid
Supplier: MP Biomedicals
Warning
Synonyms: (1S,3aR,4S,8aS)-4,8,8-Trimethyl-9-methylenedecahydro-1,4-methanoazulene, (+)-Longifolene
0215526501 0215526505
ICNA0215526501EA 53.6 EUR
ICNA0215526501 ICNA0215526505
Longifolene, clear, colourless liquid
Longifolene
(+)-Longifolene is a building block for the synthesis of hydroazulenes. It is also used in the preparation of dilongifolylborane, an effective chiral hydroborating agent.
Soluble in benzene; insoluble in water.
Formula: C₁₅H₂₄
MW: 204.357 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00082306
CAS Number: 475-20-7