Lactobionic acid 97, Sigma-Aldrich®

Supplier: Merck

Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid

MQ200

153516-100G 153516-25G
SIAL153516-100GEA 375 EUR
SIAL153516-100G SIAL153516-25G
Lactobionic acid 97, Sigma-Aldrich®
Lactobionic acid
Formula: C₁₂H₂₂O₁₂
MW: 358.30 g/mol
MDL Number: MFCD00078147
CAS Number: 96-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR