Anti-CYPs Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADIMFO100D ADIMFO100F
ENZOADIMFO100DEA 227 EUR
ENZOADIMFO100D ENZOADIMFO100F
Anti-CYPs Rabbit Polyclonal Antibody
Antibodies
Cytochrome p450 is a highly conserved and diverse family of heme-containing proteins that function in the metabolism of numerous endogenous and exogenous substrates.

  • Dilution range: Western blot (1:1,000)
  • Detects a band of ~50 - 55 kDa by Western blot

Type: Primary
Antigen: CYPs
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Clam, Dog, Guinea pig, Horse, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR